
1,2,3,4-Tetrahydro-2,2,4,7-tetramethylquinoline 97, Sigma-Aldrich®
:59388-58-8
:
:C₁₃H₁₉N
:MFCD00799587
:189.30 g/mol
:123 °C/5 mmHg (lit.)
:0,975 g/mL (25 °C) (lit.)
:M-Clarity™ (Niveau de qualité) = MQ100
:113 °C (coupe fermée)
:GHS07
:Room Temperature
- Appearance (Colour)
May Darken In Storage:Colorless to Yellow to Brown - Appearance (Form):Liquid
- Infrared spectrum:Conforms to Structure
- Purity (Titration by HCL04):96.5 - 103.5 %
- Purity (GC):≥96.5%
Frequently Bought Together