Order Entry
Belgium
ContactUsLinkComponent
 
Toxic
Health
Corrosive
Osmium tetraoxide solution, Sigma-Aldrich®
Catalog # 75633-2ML
CAS Number:  
Toxic
Health
Corrosive
Osmium tetraoxide solution, Sigma-Aldrich®
Catalog # 75633-2ML
CAS Number:  

Specifications

  • Pk:
    2 mL
  • Seq:
    2145

Specifications

About this item

 :
 :254.23 g/mol
Density:1.0 g/mL (20 °C)
Storage Temperature:Refrigerator
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS05
CAS Number:20816-12-0
Formula:O₄Os
MDL Number:MFCD00011150