Order Entry
Belgium
ContactUsLinkComponent
 
Alcian blue 8GX 1% in 3% acetic acid for microscopy, Sigma-Aldrich®
 
Alcian blue 8GX 1% in 3% acetic acid for microscopy, Sigma-Aldrich®

 

  • Appearance (Colour):
    Blue to Very Dark Blue Blue/Blue-Green to Very Deep Blue/Blue-Green
  • Appearance (Form):
    Liquid
  • Density (D20/4):
    0.996 - 1.016
  • Refractive index at 20 °C:
    1.331 - 1.341
  • Wavelength (1)(UV):
    605 - 625 nm
  • Extinction (1) (UV):
    (1%/1 cm)
  • Solvent (UV):
    Water

 

Frequently Bought Together