Order Entry
Belgium
ContactUsLinkComponent
Azure A eosinate, powder, crystals or chunks, Sigma-Aldrich®
Azure A eosinate, powder, crystals or chunks, Sigma-Aldrich®
Catalog # 200212-25G
CAS Number:  
undefined
Azure A eosinate, powder, crystals or chunks, Sigma-Aldrich®
Catalog # 200212-25G
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance (Form):
    Powder or Crystals
  • Infrared spectrum:
    Conforms to Structure
  • Wavelength:
    625 - 631 nm
  • Absorbance:
    ≥0.4
  • Absorbance, c = 0.005 g/l; In Methanol:
    ≥0.4

Specification Test Results

About this item

 :
Storage Temperature:Ambient
EINECS:263-491-2
CAS Number:62298-43-5
MDL Number:MFCD00012112