Dodecamolybdophosphoric acid hydrate solution ready-to-use spray and plunge reagent for chromatography, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:51429-74-4
:
Formula:H₃[P(Mo₃O₁₀)₄]·nH₂O
MDL Number:MFCD00149913
:1843.27 g/mol
ADR:3,II
:GHS02
:GHS05
:GHS07
Frequently Bought Together