Specifications
- Pk:250 mL
- Seq:2590
- Environmentally Preferable:
Specifications
About this item
Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
:
:319.86 g/mol
CAS Number:61-73-4
:MFCD00012111
Formula:C₁₆H₁₈ClN₃S
MDL Number:MFCD00150008