Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
p-Anisaldehyde
p-Anisaldehyde solution, spray reagent, Sigma-Aldrich®
p-Anisaldehyde solution, spray reagent, Sigma-Aldrich®
p-Anisaldehyde solution, spray reagent, Sigma-Aldrich®
SRA1-1KT
:
SIGMA ALDRICH MICROSCOPY
:
p-Anisaldehyde solution, spray reagent, Sigma-Aldrich®
SRA1-1KT
:
SIGMA ALDRICH MICROSCOPY
:
Pk:
1 KIT
Seq:
0075
:
:
9,II
:
136.15 g/mol
:
GHS02
:
GHS06
:
GHS08
:
Ambient
:
123-11-5
:
C₈H₈O₂
:
MFCD00003385