Order Entry
Belgium
ContactUsLinkComponent
Tetramethylrhodamine isothiocyanate Isomer R (6-TRITC isomer R), Sigma-Aldrich®
Tetramethylrhodamine isothiocyanate Isomer R (6-TRITC isomer R), Sigma-Aldrich®
Certificates
undefined
Tetramethylrhodamine isothiocyanate Isomer R (6-TRITC isomer R), Sigma-Aldrich®

Specification Test Results

  • Appearance (Form):
    Powder
  • Solubility (Colour):
    Faint Red to Very Dark Red to Very Dark Purple
  • Solubility (Form)
    10 mg/mL , 1M NH4OH in MeOH:
    Solution
  • 1H NMR Spectrum:
    Conforms to Structure
  • Solvent Content (by GC):
    ≤4 %
  • Nitrogen:
    ≥ 7.5 %
  • Dye Content:
    ≥80 %
  • Labeling Efficiency:
    ≥50 %

Specification Test Results

Frequently Bought Together