Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Silver enhancer solution A
Silver enhancer solution A, Sigma-Aldrich®
Silver enhancer solution A, Sigma-Aldrich®
Environment
Silver enhancer solution A, Sigma-Aldrich®
Catalog # S5020-100ML
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Expand image
Environment
Silver enhancer solution A, Sigma-Aldrich®
Catalog # S5020-100ML
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
Pk:
100 mL
Seq:
2440
Specifications
About this item
Product Details
:
:
GHS09
Storage Temperature:
Refrigerator
Product Details