Order Entry
Belgium
ContactUsLinkComponent
p-Amidinophenyl p-(6-amidino-2-indolyl)phenyl ether dihydrochloride, Sigma-Aldrich®
p-Amidinophenyl p-(6-amidino-2-indolyl)phenyl ether dihydrochloride, Sigma-Aldrich®
Catalog # A8675-5MG
CAS Number:  
undefined
p-Amidinophenyl p-(6-amidino-2-indolyl)phenyl ether dihydrochloride, Sigma-Aldrich®
Catalog # A8675-5MG
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance (Colour):
    Yellow-Brown to Brown
  • Appearance (Form):
    Powder
  • Solubility (Colour):
    Yellow to Orange
  • Solubility (Turbidity) 1 mg plus 0.1 ml H₂O:
    Clear
  • Carbon:
    50.8 - 60.8 %
  • Nitrogen:
    13.5 - 17.0 %
  • Dye Content, Based on Nitrogen content (approx 90%):
    ≥85 %

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
 :442.34 g/mol
Storage Temperature:Room Temperature
CAS Number:55453-00-4
Formula:C₂₂H₁₉N₅O
MDL Number:MFCD00078924