Order Entry
Belgium
ContactUsLinkComponent
 
Alcian blue solution, Sigma-Aldrich®
 
Alcian blue solution, Sigma-Aldrich®
 

 

  • Appearance (Colour):
    Dark Blue to Very Dark Blue
  • Appearance (Form)
    Sediment free:
    Liquid
  • Absorbance
    1:400 in H2O:
    0.45 - 0.60 Abs UN
  • pH:
    2.4 - 2.6

 

Frequently Bought Together