
Fast Garnet GBC sulphate salt ≥90% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:101-89-3
:
Formula:C₁₄H₁₄N₄O₄S
MDL Number:MFCD00011883
:334.35 g/mol
:M-Clarity™ quality level = MQ200
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance (Colour):Orange to Very Dark Orange and Orange-Brown to Very Dark Orange-Brown and Red to Very Dark
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Solubility (Turbidity)
5mg/ml, H2O:Clear - Solubility (Colour):Red to Very Dark Red and Red-Brown to Very Dark Red Brown
- Dye Content Calculated:≥90.0 %
Specification Test Results
Frequently Bought Together