


Giemsa's stain modified, Sigma-Aldrich®
Catalog # GS1L-1L
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
ADR:3,II
:GHS02
:GHS06
:GHS08
Storage Temperature:Ambient
:1
CAS Number:51811-82-6
MDL Number:MFCD00081642