Order Entry
Belgium
ContactUsLinkComponent
Ethidium monoazide bromide (EMA) ≥95% (by HPLC), Sigma-Aldrich®
Ethidium monoazide bromide (EMA) ≥95% (by HPLC), Sigma-Aldrich®
Catalog # E2028-5MG
CAS Number:  
undefined
Ethidium monoazide bromide (EMA) ≥95% (by HPLC), Sigma-Aldrich®
Catalog # E2028-5MG
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :420.305 g/mol
Storage Temperature:Freezer
CAS Number:58880-05-0
Formula:C₂₁H₁₈BrN₅