Order Entry
Belgium
ContactUsLinkComponent
Formvar solution ~1% in 1,2-dichloroethane for microscopy, Sigma-Aldrich®
Flammable
Toxic
Health
Formvar solution ~1% in 1,2-dichloroethane for microscopy, Sigma-Aldrich®
Catalog # 09823-100ML
CAS Number:  
Flammable
Toxic
Health
Formvar solution ~1% in 1,2-dichloroethane for microscopy, Sigma-Aldrich®
Catalog # 09823-100ML
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance (colour):
    Colourless to almost colourless
  • Appearance (form):
    Liquid
  • Density D20/4:
    1.220 - 1.256
  • Refractive index N20/D:
    1.443 - 1.449

Specification Test Results

About this item

 :
Density:1,220…1,256 g/ml (20 °C)
Flash Pt:16 °C (closed cup)
ADR:3,II
 :M-Clarity™ quality level = MQ100
 :GHS02
 :GHS06
 :GHS08
CAS Number:63148-64-1
Formula:(C₃H₆O₂)n
MDL Number:MFCD00131103