Order Entry
Belgium
Orders LinkContactUsLinkComponent
Citrate Buffer, Concentrate, Sigma-Aldrich®

Citrate Buffer, Concentrate, Sigma-Aldrich®

Citrate concentrate, 380 mmol/L, 5,4 (when diluted according to procedure 386A), 20 mL
Catalog # 3861-20ML
Citrate Buffer, Concentrate, Sigma-Aldrich®
Catalog # 3861-20ML
CAS Number:  
Citrate Buffer, Concentrate, Sigma-Aldrich®

Citrate concentrate, 380 mmol/L, 5,4 (when diluted according to procedure 386A), 20 mL

CAS Number:  
Price
133,00 €
Log in for availability
1 * 20 mL
Product Details & Documents

Citrate buffer solution, 0.38 mol/L, pH 5.4 when diluted according to procedure 386A.

  • For in vitro diagnostic use

Documents

Specifications

Specifications

  • Concentration:
    380 mmol/L
  • Pk:
    20 mL
  • Description:
    Citrate concentrate
  • pH value:
    5,4 (when diluted according to procedure 386A)
  • Cat. No.:
    3861-20ML
  • M-Clarity quality level:
    MQ500
  • Storage temperature:
    2 - 8°C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.