Orders LinkContactUsLinkComponent
DNAOUT™ DNA Removal Solution
DNAOUT™ DNA Removal Solution
Supplier:   G-Biosciences
DNAOUT™ DNA Removal Solution
Supplier:  G-Biosciences
Certificates
DNAOUT™ DNA Removal Solution
DNAOUT™ DNA Removal Solution
2 Options Available Below
Product Details & Documents
DNAOUT™ is a DNA removal solution for the rapid decontamination of work areas from contaminating DNA. Simply spray the work area and wait 10-15 seconds and then rinse with Molecular Grade Water.
  • Ready-to-use with convenient spray bottle and refill bottles available
  • Remove contaminating DNA prior to sensitive work, including PCR
  • Removes amplicon, plasmid and genomic DNA
  • Fast acting (10-15 seconds)
  • Ideal for cleaning centrifuge tubes, reaction tubes, PCR machine surface, pipettes, lab benches, lab equipment and more
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 786-74
    DNA OUT™, spray bottle
    DNA OUT™, spray bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 58,40 €
    58,40 €
    1 * 250 mL
       58,40 €
  • 786-75
    DNA OUT™, refill
    DNA OUT™, refill
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 113,00 €
    113,00 €
    1 * 1 L
       113,00 €