ContactUsLinkComponent
Antimicrobial foam soap, Tork
  SCAA520800
 :  Essity
Antimicrobial foam soap, Tork
  SCAA520800
 :  Essity
  • Description:
    Tork antimicrobial foam soap (Biocide), 1 L
  • Pk:
    6
Antimicrobial foam soap made of low-alcohol formula is very effective against bacteria, viruses and yeasts. It combines normal hand cleaning with disinfection on one procedure. In addition, no perfume or dyes are incorporated and allergic reactions are thus reduced.
Look: white foam
Color: Transparent
Smell: Alchohol Smell
 : Certified with EN 1499.
 : Use biocides safely. Always read the label and product information before use.