ContactUsLinkComponent
Liquid sanitiser, divosan hypochlorite VT3
Liquid sanitiser, divosan hypochlorite VT3
Catalog # DIVY100886807
Supplier:  Diversey
Liquid sanitiser, divosan hypochlorite VT3
Catalog # DIVY100886807
Supplier:  Diversey
Specifications
  • Description:
    Divosan hypochlorite VT3, liquid sanitiser
  • Packed:
    20 L
  • Pk:
    20 L
Specifications
About this item
Divosan Hypochlorite VT3 is a concentrated liquid sanitiser for general use. It has a wide antibacterial spectrum and is effective against most vegetative forms of micro-organisms.
  • Cost efficient and highly effective sanitiser
  • Excellent stain removing properties
  • Non-foaming and free-rinsing
  • Effective in soft or hard water
  • Powerful oxidising action that assists stain removal
Can be used as a final step sanitiser in both open and closed plant (CIP) systems. Suitable for manual, soak and spray applications in open plant cleaning and also for automatic dosing for CIP.
Applicable in healthcare, lodging, food service, retail, government, food processing.
Caution: Use biocides safely. Always read the label and product information before use.