Disinfectant detergent, Suma Tab D4
Catalog # DIVY100889121
Supplier: Diversey
Specifications
- Description:Disinfectant detergent, Suma Tab D4
- pH:6.0
- Packed:4×300 pieces
- Pk:4 SET
Specifications
About this item
Suma Tab D4 is a fast acting chlorine tablet, which allows cleaning and disinfection of all surfaces in the food environment in a single operation.
- Effective on a wide spectrum of microorganisms, improving the level of hygiene
- Effective at low temperature
- Stabilised chlorine tablet
- Longer shelf life
- Non-corrosive, safe on all surfaces
- Fast-acting
These are fast acting, thus, effective at low temperatures. These are also suitable for disinfecting vegetables.
Caution: Use biocides safely. Always read the label and product information before use.