Order Entry
Belgium
ContactUsLinkComponent
Fluorinert™ FC-40, Sigma-Aldrich®
Fluorinert™ FC-40, Sigma-Aldrich®
Catalog # F9755-250ML
CAS Number:  
Fluorinert™ FC-40, Sigma-Aldrich®
Catalog # F9755-250ML
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
CAS Number:86508-42-1
 :311-89-7
MDL Number:MFCD00131095