ContactUsLinkComponent
Molecular detection assay 2 - Campylobacter
Molecular detection assay 2 - Campylobacter
Catalog # MMMPMDA2CAM96
Supplier:  3M Food Safety
undefined
Molecular detection assay 2 - Campylobacter
Catalog # MMMPMDA2CAM96
Supplier:  3M Food Safety
Specifications
  • Description:
    Campylobacter 96 tests/kit
  • Pk:
    96 Tests
Specifications
About this item
3M™ Molecular Detection Assay 2 - Campylobacter is used with the 3M™ Molecular Detection System for the rapid and specific detection of Campylobacter in enriched food and environmental samples.
  • One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees
  • Use with CE250 3M™ Campylobacter enrichment broth for fast and reliable results
  • Ready-to-use and pre-dispensed reagents provide convenience and minimise the chance of human error
  • Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification
  • The ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently
  • Real-time amplification and detection process identifies positive results in as early as 15 minutes and allows you to make critical decisions faster
As with all test methods, the source, formulation and quality of enrichment medium can influence the results. 3M has developed a Campylobacter Enrichment Broth, CE250, to be used with the 3M Molecular Detection Assay 2 Campylobacter.