ContactUsLinkComponent
Enviro swab
Enviro swab
Catalog # MMMPENVSWB100
Supplier:  3M Food Safety
undefined
Enviro swab
Catalog # MMMPENVSWB100
Supplier:  3M Food Safety
Specifications
  • Description:
    3M™ Enviro Swab
  • Pk:
    100
Specifications
About this item
The 3M™ Enviro Swab is a pre-moistened swab, ready to use for collecting environmental samples on all types of food contact surfaces in the food industry.
  • Ready-to-use swab reaches into corners and hard-to-reach areas for increased accuracy
  • Tamper-evident band indicates if the swab has been used
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant
  • Resealable tube provides tight closure for sample transportation
  • Self-contained swab and vial for increased productivity and efficiency
Each swab is in a close sealed tube and pre-moistened with a neutralising solution, avoiding the need to moisten the swabs and reducing the risk of cross contamination. The large swab head and shaped tip collect organisms from hard to reach areas efficiently. The tamper-evident band acts as a good safety indicator to show whether the swab has been used. Resealable tubes prevent one swab from contaminating other testing swabs making it ideal for use throughout the plant. The 3M™ Enviro Swab is not recommended for use in 'dry' facilities where completely dry sampling is required. The Hazard Analysis Critical Control Point (HACCP) approach is the most widely accepted food safety management protocol worldwide. Sample collection products from 3M are a crucial part of this approach to identify potential food-borne hazards and pinpoint where they reside. Portability, stable enrichment media, and easy-to-handle swabs and sponges are designed to test for a wide variety of potential hazards and deliver exceptionally accurate results. They are engineered to safely gather, hold, and secure contents from the collection site to the lab without leaking or compromising the sample.