Order Entry
Belgium
Orders LinkContactUsLinkComponent
Aqueous mounting medium, Fluoroshield™ with DAPI, Sigma-Aldrich®

Aqueous mounting medium, Fluoroshield™ with DAPI, Sigma-Aldrich®

Aqueous mounting medium with DAPI, 20 mL
Catalog # F6057-20ML
Aqueous mounting medium, Fluoroshield™ with DAPI, Sigma-Aldrich®
Catalog # F6057-20ML
CAS Number:  
Aqueous mounting medium, Fluoroshield™ with DAPI, Sigma-Aldrich®

Aqueous mounting medium with DAPI, 20 mL

CAS Number:  
Price
120,00 €
Log in for availability
1 * 20 mL
Product Details & Documents

Fluoroshield™ with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.

  • Storage temperature: 2 - 8 °C

This mounting medium is fortified with DAPI which is a counterstain for DNA. For use in in situ hybridization techniques or other methods where fluorescence of DNA staining is required. DAPI excites at 360 nm and emits at 460 nm, producing a blue fluorescence. RNA is also stained with DAPI.

Applications:
- Counterstain in immunocytochemistry to label nuclei of neural progenitor cells
- Histology mounting medium for cardiac tissues sections for immunofluorescence studies
- Mounting solution for immunostained heart sections for nuclei detection

Specifications

Specifications

  • Pk:
    20 mL
  • Description:
    Aqueous mounting medium with DAPI

Specifications