Order Entry
Belgium
ContactUsLinkComponent
Aqueous mounting medium, Fluoroshield™ with 1,4-Diazabicyclo[2.2.2]octane, Sigma-Aldrich®
Aqueous mounting medium, Fluoroshield™ with 1,4-Diazabicyclo[2.2.2]octane, Sigma-Aldrich®
Catalog # F6937-20ML
CAS Number:  
Aqueous mounting medium, Fluoroshield™ with 1,4-Diazabicyclo[2.2.2]octane, Sigma-Aldrich®
Catalog # F6937-20ML
CAS Number:  

Specifications

  • Pk:
    20 mL
  • Description:
    Aqueous mounting medium with 1,4-Diazabicyclo[2.2.2]octane

Specifications

About this item

Fluoroshield™ with 1,4-Diazabicyclo[2.2.2]octane is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.

  • Storage temperature: 2 - 8 °C

Fluoroshield™ prevents rapid photobleaching of FITC, Texas Red®, AMCA, Alexa Fluor™ 488, Alexa Fluor 594, Cy® dyes, and tetramethylrhodamine. This mounting medium is fortified with 1,4-Diazabicyclo[2.2.2]octane (a generic version of DABCO™), an anti-fade reagent used to retard photobleaching of fluorescein and other fluorescent dyes.