Order Entry
Belgium
Orders LinkContactUsLinkComponent
Trizmal™ Buffer, Sigma-Aldrich®

Trizmal™ Buffer, Sigma-Aldrich®

Trizmal™ Buffer, Sigma-Aldrich®
Certificates

2 Options Available Below

Product Details & Documents

For use in the Naphthol AS-D Chloroacetate Esterase Kit No. 91-C.

  • Trizma® maleate buffer

Caution: Trizmal is a trademark of Sigma-Aldrich Co. LLC

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 903C-50ML
  • 913-50ML
  • Storage temperature:
  • RT
  • 2 - 8 °C
  • M-Clarity quality level:
  • MQ500
  • MQ500

Specifications

Product Family Options

Product Information

  • pH value
    Pk
    Availability
    Price
  • 903C-50ML
    Trizmal™ buffer, 0,2 M
    Trizmal™ buffer, 0,2 M
    6,3 (concentrate)
    50 mL
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - 123,00 €
    123,00 €
    1 * 50 mL
       123,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    903C-50ML
    Storage temperature
    RT
    M-Clarity quality level
    MQ500
  • 913-50ML
    Trizmal™ buffer, 1,0 M
    Trizmal™ buffer, 1,0 M
    6,3 (concentrate)
    50 mL
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - 106,00 €
    106,00 €
    1 * 50 mL
       106,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    913-50ML
    Storage temperature
    2 - 8 °C
    M-Clarity quality level
    MQ500
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.