Order Entry
Belgium
ContactUsLinkComponent
Peroxidase (Myeloperoxidase) Leukocyte Kit Hanker-Yates substrate kit, Sigma-Aldrich®
Toxic
Environment
Peroxidase (Myeloperoxidase) Leukocyte Kit Hanker-Yates substrate kit, Sigma-Aldrich®
Catalog # 390A-1KT
CAS Number:  
Toxic
Environment
Peroxidase (Myeloperoxidase) Leukocyte Kit Hanker-Yates substrate kit, Sigma-Aldrich®
Catalog # 390A-1KT
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ500 (more info.)
UN:3316
 :GHS06
 :GHS09
Storage Temperature:Refrigerator
 :1
MDL Number:MFCD00131641