Orthophosphoric acid 85+% for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7664-38-2
:
Formula:H₃PO₄
MDL Number:MFCD00011340
:98 g/mol
Melting Pt:42 °C
Storage Temperature:Ambient
UN:3453
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Clear colorless Viscous liquid
- Density:1.70 kg/l (about)
- UV:(1 M solution)
- Gradient analysis:pass test
Specification Test Results
Frequently Bought Together