Order Entry
Belgium
ContactUsLinkComponent
Potassium hydrogen phthalate for HPLC
Potassium hydrogen phthalate for HPLC
Certificates
undefined
Potassium hydrogen phthalate for HPLC

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (10 % in water) Clear colorless
  • UV:
    (10 % in water)
  • pH:
    3.5 to 5.0 (5 % in water)
  • Titration with NaOH:
    99.4 to 100.6 %

Specification Test Results

Frequently Bought Together