ContactUsLinkComponent
Controlled-rate freezers, CryoMed
Controlled-rate freezers, CryoMed
Certificates
undefined
Controlled-rate freezers, CryoMed
Limited Time Sale
Select items below are currently on sale
Specifications
  • Catalog No:
  • 471-1248
  • 471-1359
  • 471-1314
  • 471-1313
  • 471-1250
  • 471-1249
  • Model:
  • CryoMed CRF 17
  • CryoMed CRF 48
  • CryoMed CRF 34
    with thermal printer
  • CryoMed CRF 17
    with thermal printer
  • CryoMed CRF 48
  • CryoMed CRF 34
  • Display:
  • Touch-screen LCD
  • Touch-screen LCD
  • Touch-screen LCD
  • Touch-screen LCD
  • Touch-screen LCD
  • Touch-screen LCD
  • Alarm:
  • Audible and visual
  • Audible and visual
  • Audible and visual
  • Audible and visual
  • Audible and visual
  • Audible and visual
  • Electrical:
  • 220 V, 50 - 60 Hz / 7FLA
  • 220 V, 50 - 60 Hz / 7FLA
  • 220 V, 50 - 60 Hz / 7FLA
  • 220 V, 50 - 60 Hz / 7FLA
  • 220 V, 50 - 60 Hz / 7FLA
  • 220 V, 50 - 60 Hz / 7FLA
  • Door type:
  • Solid
  • Solid
  • Solid
  • Solid
  • Solid
  • Solid
  • Door lock:
  • via access code
  • via access code
  • via access code
  • via access code
  • via access code
  • via access code
  • Weight:
  • 69,8 kg
  • 87,1 kg
  • 79,3 kg
  • 70,3 kg
  • 86,6 kg
  • 78,9 kg