Sampling spoon, long handle, disposable Bio
About this item
The sampling spoon with long handle can be used to remove powders, granulates, pastes or liquids. The long handle makes it easy to reach the bottoms of larger containers as well and allows for sampling of drums, jerrycans and bottles with small opening diameters. Thanks to the indentations on the handle, the disposable spoon rests comfortably and securely in the hand.
- Product made from renewable raw materials
- Bio-PE, white
- Individually packaged and optionally sterilised
- Handle length 300 mm
- For single use
- Clean room manufactured (clean room class 7)
- Complies with EU food requirements and FDA regulations
The laboratory spoon is suitable for single use in the sensitive areas of pharmaceuticals, chemistry, cosmetics, animal feed and food and beverage.
4 Options Available Below
Product Details & Documents
The sampling spoon with long handle can be used to remove powders, granulates, pastes or liquids. The long handle makes it easy to reach the bottoms of larger containers as well and allows for sampling of drums, jerrycans and bottles with small opening diameters. Thanks to the indentations on the handle, the disposable spoon rests comfortably and securely in the hand.
- Product made from renewable raw materials
- Bio-PE, white
- Individually packaged and optionally sterilised
- Handle length 300 mm
- For single use
- Clean room manufactured (clean room class 7)
- Complies with EU food requirements and FDA regulations
The laboratory spoon is suitable for single use in the sensitive areas of pharmaceuticals, chemistry, cosmetics, animal feed and food and beverage.
Specifications for Product Family
Specifications
- Catalog No:
- 331-0328
- 331-0327
- 331-0330
- 331-0329
- Environmentally Preferable:
- Low Manufacturing Impact|Sustainable Materials
- Low Manufacturing Impact|Sustainable Materials
- Low Manufacturing Impact|Sustainable Materials
- Low Manufacturing Impact|Sustainable Materials
- Environmental certification:
- ACT-Label
- ACT-Label
- ACT-Label
- ACT-Label
Specifications
Product Family Options
Product Information
- LengthCapacityMaterialAvailabilityPrice
- 331-0328Sampling spoon, non sterileSpecifications:
Low Manufacturing Impact|Sustainable Materials More
Specifications
Cat. No.331-0328Environmentally PreferableLow Manufacturing Impact|Sustainable MaterialsEnvironmental certificationACT-LabelLow Manufacturing Impact|Sustainable Materials - 331-0327Sampling spoon, sterileSpecifications:
Low Manufacturing Impact|Sustainable Materials More
Specifications
Cat. No.331-0327Environmentally PreferableLow Manufacturing Impact|Sustainable MaterialsEnvironmental certificationACT-LabelLow Manufacturing Impact|Sustainable Materials - 331-0330Sampling spoon, non sterileSpecifications:
Low Manufacturing Impact|Sustainable Materials More
Specifications
Cat. No.331-0330Environmentally PreferableLow Manufacturing Impact|Sustainable MaterialsEnvironmental certificationACT-LabelLow Manufacturing Impact|Sustainable Materials - 331-0329Sampling spoon, sterileSpecifications:
Low Manufacturing Impact|Sustainable Materials More
Specifications
Cat. No.331-0329Environmentally PreferableLow Manufacturing Impact|Sustainable MaterialsEnvironmental certificationACT-LabelLow Manufacturing Impact|Sustainable Materials