Orders LinkContactUsLinkComponent
CryoStorage vials
CryoStorage vials
Supplier:   EPPENDORF
CryoStorage vials
Supplier:  EPPENDORF
Certificates
CryoStorage vials
CryoStorage vials
10 Options Available Below
Product Details & Documents
CryoStorage vials for storage at −80 °C are labeled with a SafeCode barcode for safe sample identification. Different volumes are available.
  • Free of DNase, RNase, human DNA, endotoxin
  • External thread, pre-capped grey
  • With 2D SafeCode (DataMatrix/ECC200), linear and human readable code
Eppendorf CryoStorage vials take advantage of the 3-level coding of the SafeCode system (datamatrix code, barcode, and clear writing) to enable safe sample identification. Up to 96 pre-labelled vials are pre-racked and pre-capped for convenient usage. The vial range covers 0,5 to 4,0 ml for a broad range of storage applications at e.g. −80 °C in the ULT freezer. These vials are the solution for modern high-density storage with highest sample safety, management, and tracking needs.
Packaging: Individually packed racks.
Product Family Options
Product Information
  • Capacity
    Cap included
    Sterile
    Packed
    Availability
    Price
  • EPPE0030079.400
    CryoStorage vial
    CryoStorage vial
    0,50 ml
    Yes
    -
    10 racks of 96 vials
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 725,10 €
    725,10 €
    1 * 960 ST
       725,10 €
  • EPPE0030079.418
    CryoStorage vial
    CryoStorage vial
    1,00 ml
    Yes
    -
    10 racks of 96 vials
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 753,90 €
    753,90 €
    1 * 960 ST
       753,90 €
  • EPPE0030079.426
    CryoStorage vial
    CryoStorage vial
    1,50 ml
    Yes
    -
    10 racks of 48 vials
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 498,20 €
    498,20 €
    1 * 480 ST
       498,20 €
  • EPPE0030079.434
    CryoStorage vial
    CryoStorage vial
    2,00 ml
    Yes
    -
    10 racks of 48 vials
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 512,60 €
    512,60 €
    1 * 480 ST
       512,60 €
  • EPPE0030079.442
    CryoStorage vial
    CryoStorage vial
    4,00 ml
    Yes
    -
    10 racks of 48 vials
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 666,70 €
    666,70 €
    1 * 480 ST
       666,70 €
  • EPPE0030079.450
    CryoStorage vial
    CryoStorage vial
    0,50 ml
    Yes
    +
    10 racks of 96 vials
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 773,10 €
    773,10 €
    1 * 960 ST
       773,10 €
  • EPPE0030079.469
    CryoStorage vial
    CryoStorage vial
    1,00 ml
    Yes
    +
    10 racks of 96 vials
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 801,90 €
    801,90 €
    1 * 960 ST
       801,90 €
  • EPPE0030079.477
    CryoStorage vial
    CryoStorage vial
    1,50 ml
    Yes
    +
    10 racks of 48 vials
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 522,20 €
    522,20 €
    1 * 480 ST
       522,20 €
  • EPPE0030079.485
    CryoStorage vial
    CryoStorage vial
    2,00 ml
    Yes
    +
    10 racks of 48 vials
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 790,69 €
    790,69 €
    1 * 480 ST
       790,69 €
  • EPPE0030079.493
    CryoStorage vial
    CryoStorage vial
    4,00 ml
    Yes
    +
    10 racks of 48 vials
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 690,70 €
    690,70 €
    1 * 480 ST
       690,70 €