Order Entry
Belgium
Orders LinkContactUsLinkComponent
Air sampler, Coriolis® COMPACT

Air sampler, Coriolis® COMPACT

Supplier:   Bertin Instruments
Air sampler, Coriolis® COMPACT
Supplier:  Bertin Instruments
Certificates
Air sampler, Coriolis® COMPACT

2 Options Available Below

Product Details & Documents

Portable dry air sampler for bio contamination control.

  • Concentrates airborne particles using a dry cyclonic technique
  • Compatible with culture and molecular biology standard methods (PCR, qPCR, or NGS)
  • Easy decontamination (hydrogen peroxide)
  • Autonomy on battery of 8 hours
  • Light and compact for indoor and outdoor use

Coriolis® COMPACT is a portable biological air sampler using a dry cyclonic technique, for indoor or outdoor air bio-contamination monitoring. Its innovative dry cyclonic technology aspirates the particles with an airflow of 50 L/m and centrifuges them in a cone. The collected particles or microorganisms (bacteria, viruses, moulds) can then be recovered by rinsing with an appropriate buffer that can be analysed afterwards using microbiological techniques.

Sample types include virus, bacteria, moulds, pollens and spores.

Suitable for environmental and pollution research, pharmaceutical, food and veterinary industries, biomedical and health environments.

Accessories information: Plastic cones are used with a screw cap to facilitate the transport and preservation of samples. Sterile and non sterile cones and caps are available (ready to use doses). The Coriolis Connect app is a comprehensive mobile app, able to monitor the Coriolis Compact. Keep track of collection campaigns directly on the app and manage sampling conditions by setting up protocols. All of the functionalities of the Coriolis Compact are now accessible directly on an Android smartphone.

Specifications for Product Family

Specifications

  • Catalog No:
  • 710-2144
  • 710-2069
  • Flow rate:
  • 50 L/min
  • Particle size:
  • 0,5 - 10 µm
  • Running time:
  • 1 min to 8 h
  • W×D×H:
  • 135×130×225 mm
  • Weight:
  • 1,2 kg (with battery)

Specifications

Product Family Options

Accessories

Product Information

  • Availability
    Price
  • 710-2144
    Coriolis® COMPACT cones and air intake
    Coriolis® COMPACT cones and air intake
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 132,00 €
    132,00 €
    1 * 10 ST
       132,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    710-2144
    Flow rate
    Particle size
    Running time
    W×D×H
    Weight

Product Information

  • Availability
    Price
  • 710-2069
    Coriolis® COMPACT portable dry air sampler
    Coriolis® COMPACT portable dry air sampler
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9 120,00 €
    9 120,00 €
    1 * 1 ST
       9 120,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    710-2069
    Flow rate
    50 L/min
    Particle size
    0,5 - 10 µm
    Running time
    1 min to 8 h
    W×D×H
    135×130×225 mm
    Weight
    1,2 kg (with battery)
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Coriolis® COMPACT box shipment standard
    Coriolis® COMPACT box shipment standard
    BERTS002078CORC0A0
    Log in for availability
    1 * 1 ST - 540,00 €