Order Entry
Belgium
ContactUsLinkComponent
 
Restroom mat runner with adhesive backing, PIG®
Catalog # 115-2586
Supplier:  NEW PIG
undefined
Restroom mat runner with adhesive backing, PIG®
Catalog # 115-2586
Supplier:  NEW PIG

Specifications

  • Type:
    Polypropylene
  • Description:
    Restroom mat runner, 1 roll, 2,2 kg
  • L×W:
    610×8000 mm
  • Pk:
    1 Roll

Specifications

About this item

Adhesive-backed mat runner lets you create custom-sized restroom mats for urinals, sinks and other wet spots.

  • Unique mat runner can be easily rolled out and cut to length for continuous floor coverage with no gaps or overlaps
  • 25-foot roll is ideal for up to 10 urinals, under rows of sinks or wherever custom sizes are needed
  • Permanently infused antimicrobial technology prevents the growth of odor-causing bacteria and mold on the mats to keep restrooms cleaner and fresher smelling
  • Proprietary, full-coverage adhesive backing keeps mats in place underfoot with no shifting, curling or flipping up to stop slip and trip hazards; peels up easily
  • Mats stick to your floor and won't budge while you clean them; simply mop them with the rest of the floor
  • Durable, Hi-Trac™ surface stands up to daily cleaning and restroom traffic
  • Absorbs water and urine

EFM™ Absorbent technology captures water on contact, wicks it away and dries out quickly, Impermeable backing layers stop liquid pass-through to prevent stains and grout damage.

Release paper on the bottom of the mat should be removed prior to installation.

Certifications: Tested and certified by the National Floor Safety Institute (NFSI) as a high-traction surface which can reduce slip and fall claims by as much as 90% when used as part of a floor safety program.
Caution: Mats take on the properties of absorbed liquids - always take proper precautions when handling saturated absorbents.