
Cerium(IV) sulphate standardised solution 0.1 N in aqueous solution
Catalog # 35650.K2
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:332.24 g/mol
Merck Index:14,1990
Density:1.05
Storage Temperature:Ambient
EINECS:237-029-5
UN:3264
ADR:8,III
:GHS05
CAS Number:13590-82-4
Formula:Ce(SO₄)₂
MDL Number:MFCD00148852