Water, AR® ACS, Deionized, Macron Fine Chemicals™
Catalog # CH453-09
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Cubetainer®
- Pk:20 L
- Seq:5302
- Pk Info:1×20 L
Specifications
Specification Test Results
- ACS - Chloride (Cl):≤0.4 ppm
- ACS - Electrical Resistivity at 25°C (Mohm.cm):≥0.5
- ACS - Heavy Metals (as Pb):≤0.01 ppm
- ACS - Nitrate (NO₃):≤0.4 ppm
- ACS - Phosphate (PO₄):≤1.0 ppm
- ACS - Residue after Evaporation:≤0.0002 %
- ACS - Silicate (as SiO₂):≤0.01 ppm
- ACS - Specific Conductance at 25°C. (/uohms⁻¹ cm⁻¹):≤2.0
- ACS - Substances Reducing Permanganate:Passes Test
- ACS - Sulfate (SO₄):≤1.0 ppm
Specification Test Results
About this item
:
:18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
:Management of Change (MOC) category = NI
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332