Order Entry
Belgium
ContactUsLinkComponent
Sodium dihydrogen phosphate monohydrate 98.0-103.0% USP, Multi-Compendial, J.T.Baker®
Sodium dihydrogen phosphate monohydrate 98.0-103.0% USP, Multi-Compendial, J.T.Baker®
Catalog # CH04-07
Supplier:  Avantor
CAS Number:  
undefined
Sodium dihydrogen phosphate monohydrate 98.0-103.0% USP, Multi-Compendial, J.T.Baker®
Catalog # CH04-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187
  • Environmentally Preferable:
  • Pk Info:
    1×12 kg

Specifications

Specification Test Results

  • Appearance (White crystals or granules):
    Passes Test
  • USP - Assay (anhydrous basis):
    98.0 - 103.0 %
  • USP - Aluminum, Calcium, and Related Elements:
    Passes Test
  • USP - Arsenic (As):
    ≤ 8 ppm
  • USP - Chloride (Cl):
    ≤ 0.014 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Insoluble Substances:
    ≤ 0.2 %
  • USP - pH (1 in 20):
    4.1 - 4.5
  • USP - Sulfate (SO₄):
    ≤ 0.15 %
  • USP - Water (H₂O):
    10.0 - 15.0 %
  • EP - Identification A:
    Passes Test
  • EP - Identification B:
    Passes Test
  • EP - Identification C:
    Passes Test
  • EP - Assay (anhydrous basis):
    98.0 - 100.5 %
  • EP - Chloride (Cl):
    ≤ 200 ppm
  • EP - Sulfate (SO₄):
    ≤ 300 ppm
  • EP - Reducing Substances:
    Passes Test
  • Endotoxin Concentration (EU/g):
    ≤ 2.5
  • EP - Iron (Fe):
    ≤ 10 ppm
  • EP - Appearance of Solution:
    Passes Test
  • EP - pH:
    4.2 - 4.5
  • BP - Identification A:
    Passes Test
  • BP - Identification B:
    Passes Test
  • BP - Identification C:
    Passes Test
  • BP - Identification D:
    Passes Test
  • BP - Acidity:
    4.2 - 4.5
  • BP - Clarity and Color of Solution:
    Passes Test
  • BP - Arsenic (As):
    ≤ 2 ppm
  • BP - Iron (Fe):
    ≤ 10 ppm
  • BP - Chloride (Cl):
    ≤ 200 ppm
  • BP - Sulfate (SO₄):
    ≤ 300 ppm
  • BP - Reducing Substances:
    Passes Test
  • BP - Loss on Drying at 130°C:
    11.5 - 14.5 %
  • BP - Assay (dried basis):
    98.0 - 100.5 %
  • JPE - Assay (dried basis):
    ≥ 98.0 %
  • JPE - Identification:
    Passes Test
  • JPE - pH:
    4.1 - 4.7
  • JPE - Clarity and Color of Solution:
    Passes Test
  • JPE - Chloride (Cl):
    ≤ 0.005 %
  • JPE - Sulfate (SO4):
    ≤ 0.019 %
  • JPE - Heavy Metals:
    ≤ 10 ppm
  • JPE - Arsenic (As):
    ≤ 2 ppm
  • ChP - Assay (dried basis):
    ≥ 98.0 %
  • ChP - Identification:
    Passes Test
  • ChP - Acidity (pH):
    4.1 - 4.7
  • ChP - Clarity and color of solution:
    Passes Test
  • ChP - Chlorides:
    ≤ 0.01 %
  • ChP - Sulfates:
    ≤ 0.05 %
  • ChP - Loss on Drying:
    10.0 - 15.0 %
  • ChP - Aluminum:
    ≤ 0.01 %
  • ChP - Calcium:
    ≤ 0.01 %
  • ChP - Heavy metals:
    ≤ 0.001 %
  • ChP - Arsenic:
    ≤ 0.0002 %
  • ChP - Water insoluble substanc:
    ≤ 0.2 %
  • ChP - Reducing substances:
    Passes Test
  • ChP - Iron:
    ≤ 0.001 %

Specification Test Results

About this item

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate
 :
 :137.99 g/mol
Merck Index:13,08734
Melting Pt:100 °C
Density:2.04 g/cm³
 :Management of Change (MOC) category = HR
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13
CAS Number:10049-21-5
Formula:NaH₂PO₄·H₂O
MDL Number:MFCD00149208