Order Entry
Belgium
ContactUsLinkComponent
Glucose Test Strips, Dosatest
Glucose Test Strips, Dosatest
Catalog # 85714.601
Supplier:  VWR Chemicals
CAS Number:  
undefined
Glucose Test Strips, Dosatest
Catalog # 85714.601
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Chemical method:
    Enzymatic
  • Pk:
    100
  • Description:
    Glucose test strips
  • Tests per kit:
    100
  • Range:
    0 • 50 • 100 • 250 • 500 • 1000 • 2000  mg/L

Specifications

About this item

Semi-quantitative test strips for the determination of glucose.

  • Ideal for rapid and easy analysis of glucose directly on-site
  • No maintenance or calibration is required

Colour change: Yellow to blue-green

Delivery: Delivered as 100 test strips in a tube with instruction leaflet.