Order Entry
Belgium
ContactUsLinkComponent
Molybdenum test strips, Dosatest
Molybdenum test strips, Dosatest
Catalog # 85706.601
Supplier:  VWR Chemicals
CAS Number:  
undefined
Molybdenum test strips, Dosatest
Catalog # 85706.601
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Chemical method:
    Dithiol
  • Pk:
    100
  • Description:
    Molybdenum test strips
  • Tests per kit:
    100
  • Range:
    0 • 5 • 20 • 50 • 100 • 250  mg/L Mo⁶⁺

Specifications

About this item

Semi-quantitative test strips for the determination of molybdenum.

  • Ideal for rapid and easy analysis of molybdenum directly on-site
  • No maintenance or calibration is required

Colour change: White to green.

Delivery: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box