ContactUsLinkComponent
i7100 Vial Label Applicator
i7100 Vial Label Applicator
Catalog # BRDY151156
Supplier:  Brady
undefined
i7100 Vial Label Applicator
Catalog # BRDY151156
Supplier:  Brady
Specifications
  • Description:
    i7100 vial label applicator
  • Temperature range:
    5 - 40°C
  • Dimensions:
    120,65W×220,98L×76,2D mm
  • Pk:
    1
  • For:
    BradyPrinter i7100
Specifications
About this item
Automate vial tube identification with the Vial Label Applicator accessory that prints and applies labels on vial tubes in a few seconds.
Works only with the 300 and 600 dpi Peel models of the BradyPrinter i7100 Industrial Label Printers.
Delivery: The accessories for the wraptor printer applicator includes cleaning kits, memory cards, additional hardware, shippping materials, and stylus.