ContactUsLinkComponent
Label printers with peel function and vial label applicator, industrial thermal transfer benchtop, BradyPrinter i7100
 :  Brady
undefined
Label printers with peel function and vial label applicator, industrial thermal transfer benchtop, BradyPrinter i7100
 :  Brady
  • Catalog No:
  • BRDY307653
  • BRDY307659
  • BRDY307652
  • 116-0938
  • Connections:
  • Ethernet, I/O connection, bluetooth (optional), WiFi (optional), serial, USB
  • Ethernet, I/O connection, bluetooth (optional), WiFi (optional), serial, USB
  • Ethernet, I/O connection, bluetooth (optional), WiFi (optional), serial, USB
  • Ethernet, I/O connection, bluetooth (optional), WiFi (optional), serial, USB
  • Display:
  • Colour touchscreen
  • Colour touchscreen
  • Colour touchscreen
  • Colour touchscreen
  • Power supply:
  • AC adaptor
  • AC adaptor
  • AC adaptor
  • AC adaptor
  • Print method:
  • Transfert thermique
  • Transfert thermique
  • Transfert thermique
  • Transfert thermique
  • Software:
  • Brady Workstation, Windows based driver for 3rd party software use
  • Brady Workstation, Windows based driver for 3rd party software use
  • Brady Workstation, Windows based driver for 3rd party software use
  • Brady Workstation, Windows based driver for 3rd party software use
  • Supported OS:
  • Windows® 7, Windows® 8, Windows® 10, Windows® Server 2012, Windows® Server 2012 R2, Windows® Server 2016
  • Windows® 7, Windows® 8, Windows® 10, Windows® Server 2012, Windows® Server 2012 R2, Windows® Server 2016
  • Windows® 7, Windows® 8, Windows® 10, Windows® Server 2012, Windows® Server 2012 R2, Windows® Server 2016
  • Windows® 7, Windows® 8, Windows® 10, Windows® Server 2012, Windows® Server 2012 R2, Windows® Server 2016
  • Temperature range:
  • 5...40 °C
  • 5...40 °C
  • 5...40 °C
  • 5...40 °C
Frequently Bought Together