Order Entry
Belgium
ContactUsLinkComponent
Cyanide test strips, Dosatest
Cyanide test strips, Dosatest
Catalog # 85703.601
Supplier:  VWR Chemicals
CAS Number:  
undefined
Cyanide test strips, Dosatest
Catalog # 85703.601
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Chemical method:
    Barbituric acid derivative
  • Pk:
    100
  • Description:
    Cyanide test strips
  • Range:
    0 • 1 • 3 • 10 • 30  mg/L CN⁻

Specifications

About this item

Semi-quantitative test strips for the determination of cyanide.

  • Ideal for rapid and easy analysis of cyanide directly on-site
  • No maintenance or calibration is required

Colour change: White to violet

Delivery: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box