Order Entry
Belgium
ContactUsLinkComponent
Chlorine, sensitive test strips, Dosatest
Chlorine, sensitive test strips, Dosatest
Catalog # 85712.601
Supplier:  VWR Chemicals
CAS Number:  
undefined
Chlorine, sensitive test strips, Dosatest
Catalog # 85712.601
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Chemical method:
    Redox reaction
  • Pk:
    100
  • Description:
    Chlorine test strips
  • Tests per kit:
    100
  • Range:
    0 • 0.1 • 0.5 • 1 • 3 • 10  mg/L Cl₂

Specifications

About this item

Semi-quantitative test strips for the determination of chlorine.

  • Ideal for rapid and easy analysis of chlorine directly on-site
  • No maintenance or calibration is required

Colour change: Yellow to violet

Delivery: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box