Order Entry
Belgium
ContactUsLinkComponent
Quaternary ammonium test strips, Dosatest, VWR®
Quaternary ammonium test strips, Dosatest, VWR®
Catalog # 85711.601
Supplier:  VWR Chemicals
CAS Number:  
undefined
Quaternary ammonium test strips, Dosatest, VWR®
Catalog # 85711.601
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Chemical method:
    Mixed indicator
  • Pk:
    100
  • Description:
    QUAT test strips
  • Tests per kit:
    100
  • Range:
    0 • 10 • 25 • 50 • 100 • 250 • 500 • 1000  mg/L Benzalkonium chloride

Specifications

About this item

Semi-quantitative test strips for the determination of QUAT.

  • Ideal for rapid and easy analysis of QUAT ammonium directly on-site
  • No maintenance or calibration is required

Colour change: Yellow  to  blue-green

Delivery: Delivered as 100 test strips in a tube with instruction leaflet.