Ascorbic acid test strips, Dosatest
Ascorbic acid test strips, 0 • 50 • 100 • 200 • 300 • 500 • 700 • 1000 • 2000 mg/L Vitamin C, Phosphormolybdenum blue, 100, 100
Fournisseur: VWR Chemicals
Aller à :
Product Details & Documents
Semi-quantitative test strips for the determination of ascorbic acid.
- Ideal for rapid and easy analysis directly on-site
- No maintenance or calibration is required
Colour change: Yellow to green-blue
Delivery information: Delivered as 100 test strips in a tube with instruction leaflet.
Documents
Certificats
Rechercher par numéro de lotspec_Ascorbic Acid Test Strips, Dosatest VWR
Product specification
VWR Indicator Papers and Strips.pdf
Product Literature
Spécifications
Spécifications
- Chemical method:Phosphormolybdenum blue
- Pk:100
- Description:Ascorbic acid test strips
- Tests per kit:100
- Range:0 • 50 • 100 • 200 • 300 • 500 • 700 • 1000 • 2000 mg/L Vitamin C
Spécifications