Order Entry
Belgium
ContactUsLinkComponent
Sulphate test strips, Dosatest, VWR®
Sulphate test strips, Dosatest, VWR®
Catalog # 85708.601
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sulphate test strips, Dosatest, VWR®
Catalog # 85708.601
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Chemical method:
    Ba-thorine-complex
  • Pk:
    100
  • Description:
    Sulphate test strips
  • Tests per kit:
    100
  • Range:
    < 200 • > 400 • > 800 • > 1200 • > 1600  mg/L SO₄²⁻

Specifications

About this item

Semi-quantitative test strips for the determination of sulphate.

  • Ideal for rapid and easy analysis of sulphate directly on-site
  • No maintenance or calibration is required

Colour change: Red to yellow

Delivery: Delivered as 100 test strips in a tube with instruction leaflet.
Caution: If the sample solution has a pH value of 4 to 8, the reaction will take place without interferences. Strongly acid solutions have to be buffered with sodium acetate, and alkaline with tartaric acid.