Order Entry
Belgium
ContactUsLinkComponent
VWR®, Sulphite test strips, Dosatest
VWR®, Sulphite test strips, Dosatest
Catalog # 85699.601
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Sulphite test strips, Dosatest
Catalog # 85699.601
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Chemical method:
    Nitroprusside / Zn-hexacyanoferrate
  • Pk:
    100
  • Description:
    Sulphite test strips
  • Tests per kit:
    100
  • Range:
    0 • 10 • 25 • 50 • 100 • 250 • 500 • 1000  mg/L SO₃²⁻

Specifications

About this item

Semi-quantitative test strips for the determination of sulphite.

  • Ideal for rapid and easy analysis of sulphite directly on-site
  • No maintenance or calibration is required

Colour change: White to salmon

Delivery: Delivered as 100 test strips in a tube with instruction leaflet.
Caution: Acid solutions such as white wine have to be buffered with solid sodium acetateas the the test paper does not react in acidic media.