Nickel Test Strips, Dosatest
Catalog # 85697.601
Supplier: VWR Chemicals
CAS Number:
Specifications
- Méthode chimique:Dimethylglyoxime
- Cdt:100
- Désignation:Nickel test strips
- Tests par set:100
- Plage de mesure:0 • 10 • 25 • 50 • 100 • 250 • 500 • 1000 mg/L Ni²⁺
Specifications
About this item
Semi-quantitative test strips for the determination of nickel.
- Ideal for rapid and easy analysis of nickel directly on-site
- No maintenance or calibration is required
Colour change: White to bright red.
Delivery: Delivered as 100 test strips in a tube with instruction leaflet.
Caution: Strongly acid solutions (pH < 2) have to be buffered to a pH value of 2 to 6 using solid sodium acetate.