ThermoSafe® FreezeTherm® Pre-Qualified Dry Ice Shippers
Supplier: Sonoco Thermosafe
Certificates
About this item
FreezeTherm® dry ice shippers are pre-qualified to several external ambient temperatures to give users confidence of likely performance.
- High-quality EPS base and lid inside a strong, outer corrugated box
- Outer box has all necessary dry ice markings to simplify the shipment
FreezeTherm® dry ice shipping systems provide ready access to convenient, affordable protection for frozen products in transit. The wide range of off-the-shelf sizes means there’s a FreezeTherm® solution for every application.