Order Entry
Belgium
ContactUsLinkComponent
Dimethyl sulphoxide ≥99.5% (by GC), HiPerSolv CHROMANORM® for HPLC
Dimethyl sulphoxide ≥99.5% (by GC), HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Dimethyl sulphoxide ≥99.5% (by GC), HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Assay (by GC):
    min. 99,5  %
  • Water (KF):
    max. 0,05 %
  • Acidity:
    max. 0,0005 meq/g
  • Residue on Evaporation:
    max. 0,001 %
  • 280 nm:
    min. 60  %
  • 300 nm:
    min. 75  %
  • 320 nm:
    min. 90  %
  • 330 nm:
    min. 95  %
  • 360 nm:
    min. 98  %
  • 365 nm:
    max. 2 ppb

Specification Test Results

Frequently Bought Together